Tisk…

6-(Dimethylamino)pyridine-3-boronic acid hydrate

Dodavatel: Apollo Scientific
Varování

OR2767-1G OR2767-500MG
APOSOR2767-1GEA 1990 CZK
APOSOR2767-1G APOSOR2767-500MG
6-(Dimethylamino)pyridine-3-boronic acid hydrate
6-(Dimethylamino)pyridine-3-boronic acid hydrate
Vzorec: C₇H₁₁BN₂O₂·H₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06656059
CAS číslo: 1256355-24-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR