3-{1-[(4-Methylphenyl)sulphonyl]-1H-indol-3-yl}acrylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-{1-[(4-Methylphenyl)sulphonyl]-1H-indol-3-yl}prop-2-enoic acid, 3-(2-Carboxyethenyl)-1-[(4-methylphenyl)sulphonyl]-1H-indole
OR27556-5G
OR27556-1G
APOSOR27556-5GEA
12670
CZK
APOSOR27556-5G
APOSOR27556-1G
3-{1-[(4-Methylphenyl)sulphonyl]-1H-indol-3-yl}acrylic acid
3-{1-[(4-Methylphenyl)sulphonyl]-1H-indol-3-yl}acrylic acid
Vzorec:
C₁₈H₁₅NO₄S MW: 341,381 g/mol Bod tání: 238…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD00238942 CAS číslo: 298187-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...