5-Nitro-3-phenyl-1H-indole-2-carboxylic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Carboxy-5-nitro-3-phenyl-1H-indole
OR27538-100MG
OR27538-1G
APOSOR27538-100MGEA
1660
CZK
APOSOR27538-100MG
APOSOR27538-1G
5-Nitro-3-phenyl-1H-indole-2-carboxylic acid
5-Nitro-3-phenyl-1H-indole-2-carboxylic acid
Vzorec:
C₁₅H₁₀N₂O₄ MW: 282,251 g/mol Bod tání: 290 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00054540 CAS číslo: 14182-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...