Tisk…

N-(1-Acetyl-1H-indolin-5-yl)-4-chloro-3-nitrobenzamide

Dodavatel: Apollo Scientific
Varování

OR27504-100MG OR27504-50MG
APOSOR27504-100MGEA 5630 CZK
APOSOR27504-100MG APOSOR27504-50MG
N-(1-Acetyl-1H-indolin-5-yl)-4-chloro-3-nitrobenzamide
N-(1-Acetyl-1H-indolin-5-yl)-4-chloro-3-nitrobenzamide
Vzorec: C₁₇H₁₄ClN₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00277956
CAS číslo: 680214-07-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR