N-[2-Amino-4-(tert-butyl)phenyl]-2-(morpholin-4-yl)acetamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-(tert-Butyl)-2-[(morpholin-4-yl)acetamido]aniline, 5-(tert-Butyl)-2-[(morpholin-4-yl)acetylamino]aniline, 4-(2-{[2-Amino-4-(tert-butyl)phenyl]amino}-2-oxoethyl)morpholine
OR27203-1G
OR27203-500MG
APOSOR27203-1GEA
21490
CZK
APOSOR27203-1G
APOSOR27203-500MG
N-[2-Amino-4-(tert-butyl)phenyl]-2-(morpholin-4-yl)acetamide
N-[2-Amino-4-(tert-butyl)phenyl]-2-(morpholin-4-yl)acetamide
Vzorec:
C₁₆H₂₅N₃O₂ MW: 291,389 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02089930 CAS číslo: 287927-90-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...