2,1,3-Benzothiadiazole-5-carbohydrazide
Dodavatel: Apollo Scientific
OR26819-1G
OR26819-250MG
APOSOR26819-1GEA
8270
CZK
APOSOR26819-1G
APOSOR26819-250MG
2,1,3-Benzothiadiazole-5-carbohydrazide
2,1,3-Benzothiadiazole-5-carbohydrazide
Vzorec:
C₇H₆N₄OS MW: 194,214 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00126469 CAS číslo: 98550-17-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...