Bicyclo[2.2.1]heptan-2-ol 95%
Dodavatel: Apollo Scientific
OR26341-5G
APOSOR26341-5GEA
3410
CZK
APOSOR26341-5G
Bicyclo[2.2.1]heptan-2-ol 95%
Bicyclo[2.2.1]heptan-2-ol
Vzorec:
C₇H₁₂O MW: 112,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00167747 CAS číslo: 1632-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...