2-Methylthiophene-3-carboxaldehyde
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methylthiophene-3-carbaldehyde, 3-Formyl-2-methylthiophene
OR2624-1G
OR2624-250MG
APOSOR2624-1GEA
22160
CZK
APOSOR2624-1G
APOSOR2624-250MG
2-Methylthiophene-3-carboxaldehyde
2-Methylthiophene-3-carboxaldehyde
Vzorec:
C₆H₆OS MW: 126,176 g/mol Bod varu: 80,5…82 °C (10 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09864856 CAS číslo: 84815-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...