Tisk…

4-acetyl-1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR25980-1G OR25980-250MG OR25980-5G
APOSOR25980-1GEA 705 CZK
APOSOR25980-1G APOSOR25980-250MG APOSOR25980-5G
4-acetyl-1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carboxylic acid
4-acetyl-1-cyclopropyl-2,5-dimethyl-1H-pyrrole-3-carboxylic acid
Vzorec: C₁₂H₁₅NO₃
MW: 221,252 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03407365
CAS číslo: 423769-78-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR