Tisk…

5-Bromo-N-ethyl-3,4-dinitrothiophen-2-amine

Dodavatel: Apollo Scientific
Nebezpečí

OR25851-100MG OR25851-250MG OR25851-50MG
APOSOR25851-100MGEA 3110 CZK
APOSOR25851-100MG APOSOR25851-250MG APOSOR25851-50MG
5-Bromo-N-ethyl-3,4-dinitrothiophen-2-amine
5-Bromo-N-ethyl-3,4-dinitrothiophen-2-amine
Vzorec: C₆H₆BrN₃O₄S
MW: 296,099 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00103270
CAS číslo: 680212-43-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR