3,4-Dichloro-N-[(diphenylphosphoryl)(fur-2-yl)methyl]aniline
Dodavatel: Apollo Scientific
|
Varování
|
OR25806-1G
OR25806-250MG
APOSOR25806-1GEA
7340
CZK
APOSOR25806-1G
APOSOR25806-250MG
3,4-Dichloro-N-[(diphenylphosphoryl)(fur-2-yl)methyl]aniline
3,4-Dichloro-N-[(diphenylphosphoryl)(fur-2-yl)methyl]aniline
Vzorec:
C₂₃H₁₈Cl₂NO₂P Storage Temperature: Ambient |
MDL Number:
MFCD00123332 CAS číslo: 680212-11-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...