Tisk…

3,5-Dimethyl-N-(2-{[(fur-2-yl)methyl]thio}ethyl)isoxazole-4-sulphonamide

Dodavatel: Apollo Scientific
Varování

OR25746-250MG OR25746-50MG
APOSOR25746-250MGEA 4300 CZK
APOSOR25746-250MG APOSOR25746-50MG
3,5-Dimethyl-N-(2-{[(fur-2-yl)methyl]thio}ethyl)isoxazole-4-sulphonamide
3,5-Dimethyl-N-(2-{[(fur-2-yl)methyl]thio}ethyl)isoxazole-4-sulphonamide
Vzorec: C₁₂H₁₆N₂O₄S₂
Storage Temperature: Ambient
MDL Number: MFCD01570599
CAS číslo: 680211-97-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR