Tisk…

Methyl 2-chloro-4,4-dimethyl-3-oxopentanoate 90%

Dodavatel: Apollo Scientific
Varování

OR25721-10G OR25721-1G
APOSOR25721-10GEA 16770 CZK
APOSOR25721-10G APOSOR25721-1G
Methyl 2-chloro-4,4-dimethyl-3-oxopentanoate 90%
Methyl 2-chloro-4,4-dimethyl-3-oxopentanoate
Vzorec: C₈H₁₃ClO₃
MW: 192,64 g/mol
Bod varu: 54…58 °C (0,1 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01570535
CAS číslo: 306935-33-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR