Tisk…

3-Benzyl-1-isopropyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

Dodavatel: Apollo Scientific

OR25580-1G OR25580-5G
APOSOR25580-1GEA 2480 CZK
APOSOR25580-1G APOSOR25580-5G
3-Benzyl-1-isopropyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
3-Benzyl-1-isopropyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile
Vzorec: C₁₅H₁₅N₃O₂
MW: 269,299 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00203983
CAS číslo: 175203-47-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR