Tisk…

5-Chloro-4-nitro-2,1,3-benzoxadiazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Chloro-4-nitrobenzofurazan

OR25547-1G OR25547-250MG
APOSOR25547-1GEA 7740 CZK
APOSOR25547-1G APOSOR25547-250MG
5-Chloro-4-nitro-2,1,3-benzoxadiazole
5-Chloro-4-nitro-2,1,3-benzoxadiazole
Vzorec: C₆H₂ClN₃O₃
MW: 199,551 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00174259
CAS číslo: 5714-17-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR