5-(Thien-2-ylthio)thiophene-2-sulphonamide
Dodavatel: Apollo Scientific
OR25462-1G
OR25462-5G
APOSOR25462-1GEA
2480
CZK
APOSOR25462-1G
APOSOR25462-5G
5-(Thien-2-ylthio)thiophene-2-sulphonamide
5-(Thien-2-ylthio)thiophene-2-sulphonamide
Vzorec:
C₈H₇NO₂S₄ MW: 277,407 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00068017 CAS číslo: 63033-64-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...