Tisk…

5-[(4-Chlorophenyl)thio]thiophene-2-sulphonamide

Dodavatel: Apollo Scientific

OR25339-1G OR25339-5G
APOSOR25339-1GEA 2690 CZK
APOSOR25339-1G APOSOR25339-5G
5-[(4-Chlorophenyl)thio]thiophene-2-sulphonamide
5-[(4-Chlorophenyl)thio]thiophene-2-sulphonamide
Vzorec: C₁₀H₈ClNO₂S₃
Storage Temperature: Ambient
MDL Number: MFCD00084975
CAS číslo: 63031-81-2

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR