Tisk…

3-amino-4-(isopropylsulphonyl)thiophene-2-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR25187-1G OR25187-5G
APOSOR25187-1GEA 622 CZK
APOSOR25187-1G APOSOR25187-5G
3-amino-4-(isopropylsulphonyl)thiophene-2-carboxylic acid
3-amino-4-(isopropylsulphonyl)thiophene-2-carboxylic acid
Vzorec: C₈H₁₁NO₄S₂
MW: 249,307 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067949
CAS číslo: 175201-89-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR