4-(9H-Fluoren-2-yl)-1,3-thiazol-2-amine
Dodavatel: Apollo Scientific
OR24874-1G
OR24874-5G
APOSOR24874-1GEA
2070
CZK
APOSOR24874-1G
APOSOR24874-5G
4-(9H-Fluoren-2-yl)-1,3-thiazol-2-amine
4-(9H-Fluoren-2-yl)-1,3-thiazol-2-amine
Vzorec:
C₁₆H₁₂N₂S MW: 264,345 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00806357 CAS číslo: 299438-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...