Tisk…

Methyl 3-[(2-methoxy-2-oxoethyl)thio]butanoate

Dodavatel: Apollo Scientific
Varování

OR24368-1G OR24368-5G OR24368-10G
APOSOR24368-1GEA 10820 CZK
APOSOR24368-1G APOSOR24368-5G APOSOR24368-10G
Methyl 3-[(2-methoxy-2-oxoethyl)thio]butanoate
Methyl 3-[(2-methoxy-2-oxoethyl)thio]butanoate
Vzorec: C₈H₁₄O₄S
MW: 206,259 g/mol
Bod varu: 106…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00205191
CAS číslo: 78648-41-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR