Ethyl 2-amino-4-propylpyrimidicne-5-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR24340-1G
OR24340-5G
APOSOR24340-1GEA
2070
CZK
APOSOR24340-1G
APOSOR24340-5G
Ethyl 2-amino-4-propylpyrimidicne-5-carboxylate
Ethyl 2-amino-4-propylpyrimidicne-5-carboxylate
Vzorec:
C₁₀H₁₅N₃O₂ MW: 209,245 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052079 CAS číslo: 127957-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...