Tisk…

2-[(5-Methylthieno[2,3-d]pyrimidin-4-yl)thio]aniline

Dodavatel: Apollo Scientific
Varování

OR24335-1G OR24335-250MG
APOSOR24335-1GEA 5200 CZK
APOSOR24335-1G APOSOR24335-250MG
2-[(5-Methylthieno[2,3-d]pyrimidin-4-yl)thio]aniline
2-[(5-Methylthieno[2,3-d]pyrimidin-4-yl)thio]aniline
Vzorec: C₁₃H₁₁N₃S₂
MW: 273,377 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00110570
CAS číslo: 175137-24-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR