Tisk…

Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate

Dodavatel: Apollo Scientific
Varování

OR24275-1G OR24275-250MG
APOSOR24275-1GEA 744 CZK
APOSOR24275-1G APOSOR24275-250MG
Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
Methyl 3-amino-5-(tert-butyl)thiophene-2-carboxylate
Vzorec: C₁₀H₁₅NO₂S
MW: 213,297 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00068163
CAS číslo: 175137-03-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR