2-(4-Nitrophenyl)thioacetamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-(2-Amino-2-thioxoethyl)nitrobenzene, 2-(4-Nitrophenyl)ethanethioamide
OR24109-10G
OR24109-1G
OR24109-250MG
APOSOR24109-10GEA
14720
CZK
APOSOR24109-10G
APOSOR24109-1G
APOSOR24109-250MG
2-(4-Nitrophenyl)thioacetamide
2-(4-Nitrophenyl)thioacetamide
Vzorec:
C₈H₈N₂O₂S MW: 196,226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00278445 CAS číslo: 76254-70-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...