5-(4-tert-Butylphenyl)-4-(4-isopropylphenyl)-1,2,4-triazole-3-thiol
Dodavatel: Apollo Scientific
|
Varování
|
OR2408-1G
OR2408-250MG
APOSOR2408-1GEA
23150
CZK
APOSOR2408-1G
APOSOR2408-250MG
5-(4-tert-Butylphenyl)-4-(4-isopropylphenyl)-1,2,4-triazole-3-thiol
5-(4-tert-Butylphenyl)-4-(4-isopropylphenyl)-1,2,4-triazole-3-thiol
Vzorec:
C₂₁H₂₅N₃S MW: 351,508 g/mol Bod tání: 224…227 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00207958 CAS číslo: 261761-26-6 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...