Tisk…

1-[1-(5-Methylisoxazol-3-yl)-1H-1,2,4-triazol-3-yl]acetone

Dodavatel: Apollo Scientific

OR23849-10G OR23849-1G OR23849-5G
APOSOR23849-10GEA 9860 CZK
APOSOR23849-10G APOSOR23849-1G APOSOR23849-5G
1-[1-(5-Methylisoxazol-3-yl)-1H-1,2,4-triazol-3-yl]acetone
1-[1-(5-Methylisoxazol-3-yl)-1H-1,2,4-triazol-3-yl]acetone
Vzorec: C₉H₁₀N₄O₂
Storage Temperature: Ambient
MDL Number: MFCD00052562
CAS číslo: 92658-77-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR