2-(1-Methylhydrazino)-3-nitropyridine
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR23731-50MG
OR23731-1G
OR23731-250MG
APOSOR23731-50MGEA
622
CZK
APOSOR23731-50MG
APOSOR23731-1G
APOSOR23731-250MG
2-(1-Methylhydrazino)-3-nitropyridine
2-(1-Methylhydrazino)-3-nitropyridine
Vzorec:
C₆H₈N₄O₂ MW: 168,153 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052235 CAS číslo: 30963-12-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...