4,5-Dimethyloctane
Dodavatel: Apollo Scientific
|
Varování
|
OR2373-1G
OR2373-5G
OR2373-250MG
APOSOR2373-1GEA
5290
CZK
APOSOR2373-1G
APOSOR2373-5G
APOSOR2373-250MG
4,5-Dimethyloctane
4,5-Dimethyloctane
Vzorec:
C₁₀H₂₂ MW: 142,282 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09038381 CAS číslo: 15869-96-2 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...