2-Bromo-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-[(Bromoacetyl)amino]-4-(4-chlorophenyl)-1,3-thiazole, N-(Bromoacetyl)-4-(4-chlorophenyl)-1,3-thiazol-2-amine
OR23652-1G
OR23652-250MG
APOSOR23652-1GEA
13790
CZK
APOSOR23652-1G
APOSOR23652-250MG
2-Bromo-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide
2-Bromo-N-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]acetamide
Vzorec:
C₁₁H₈BrClN₂OS MW: 331,616 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00113279 CAS číslo: 448224-88-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...