Tisk…

1-(Benzofuran-3-yl)-2-bromoethan-1-one

Dodavatel: Apollo Scientific
Nebezpečí

OR23402-1G OR23402-250MG
APOSOR23402-1GEA 2610 CZK
APOSOR23402-1G APOSOR23402-250MG
1-(Benzofuran-3-yl)-2-bromoethan-1-one
1-(Benzofuran-3-yl)-2-bromoethan-1-one
Vzorec: C₁₀H₇BrO₂
MW: 239,065 g/mol
Bod tání: 136…137 °C
Storage Temperature: Ambient
MDL Number: MFCD03407369
CAS číslo: 187657-92-7
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR