Tisk…

3-(1H-Pyrrol-1-yl)thiophene-2-carboxaldehyde

Dodavatel: Apollo Scientific

OR23356-1G OR23356-5G
APOSOR23356-1GEA 6350 CZK
APOSOR23356-1G APOSOR23356-5G
3-(1H-Pyrrol-1-yl)thiophene-2-carboxaldehyde
3-(1H-Pyrrol-1-yl)thiophene-2-carboxaldehyde
Vzorec: C₉H₇NOS
MW: 177,223 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03086144
CAS číslo: 107073-28-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR