1-[4-(Bromomethyl)phenyl]-1H-pyrazole 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
α-Bromo-4-(1H-pyrazol-1-yl)toluene, 4-(1H-Pyrazol-1-yl)benzyl bromide
OR23293-1G
OR23293-5G
APOSOR23293-1GEA
5200
CZK
APOSOR23293-1G
APOSOR23293-5G
1-[4-(Bromomethyl)phenyl]-1H-pyrazole 95%
1-[4-(Bromomethyl)phenyl]-1H-pyrazole
Vzorec:
C₁₀H₉BrN₂ MW: 237,096 g/mol Bod tání: 77…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD02681940 CAS číslo: 368869-85-6 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...