5-(4-Iodophenyl)isoxazole
Dodavatel: Apollo Scientific
|
Varování
|
OR23193-1G
OR23193-250MG
APOSOR23193-1GEA
3740
CZK
APOSOR23193-1G
APOSOR23193-250MG
5-(4-Iodophenyl)isoxazole
5-(4-Iodophenyl)isoxazole
Vzorec:
C₉H₆INO MW: 271,055 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02681898 CAS číslo: 160377-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...