Tisk…

Dioctyl malonate

Dodavatel: Apollo Scientific
Varování

Synonyms: Dioctylmalonate

OR22269-1G OR22269-250MG
APOSOR22269-1GEA 2280 CZK
APOSOR22269-1G APOSOR22269-250MG
Dioctyl malonate
Dioctyl malonate
Vzorec: C₁₉H₃₆O₄
MW: 328,487 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00048936
CAS číslo: 16958-88-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR