Tisk…

1-Bromo-4-butoxybenzene 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-Bromo-4-n-butoxylbenzene, 1-(4-Bromophenoxy)butane, 4-Bromophenyl but-1-yl ether

OR21904-1G OR21904-5G
APOSOR21904-1GEA 622 CZK
APOSOR21904-1G APOSOR21904-5G
1-Bromo-4-butoxybenzene 97%
1-Bromo-4-butoxybenzene
Vzorec: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037100
CAS číslo: 39969-57-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR