Tisk…

1,5-Dibromo-2,4-dimethoxybenzene 95%

Dodavatel: Apollo Scientific

OR2173-1G OR2173-5G
APOSOR2173-1GEA 1870 CZK
APOSOR2173-1G APOSOR2173-5G
1,5-Dibromo-2,4-dimethoxybenzene 95%
1,5-Dibromo-2,4-dimethoxybenzene
Vzorec: C₈H₈Br₂O₂
MW: 295,956 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061097
CAS číslo: 24988-36-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR