3-(trichloromethyl)-1,2,4-thiadiazol-5-amine
Dodavatel: Apollo Scientific
OR21717-1G
OR21717-250MG
OR21717-500MG
OR21717-2.5G
APOSOR21717-1GEA
14720
CZK
APOSOR21717-1G
APOSOR21717-250MG
APOSOR21717-500MG
APOSOR21717-2.5G
3-(trichloromethyl)-1,2,4-thiadiazol-5-amine
3-(trichloromethyl)-1,2,4-thiadiazol-5-amine
Vzorec:
C₃H₂Cl₃N₃S MW: 218,492 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01109217 CAS číslo: 7523-57-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...