2-[(4-Methylphenyl)thio]nicotinamide
Dodavatel: Apollo Scientific
OR21712-1G
OR21712-5G
APOSOR21712-1GEA
2280
CZK
APOSOR21712-1G
APOSOR21712-5G
2-[(4-Methylphenyl)thio]nicotinamide
2-[(4-Methylphenyl)thio]nicotinamide
Vzorec:
C₁₃H₁₂N₂OS MW: 244,312 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00052123 CAS číslo: 175135-83-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...