Tisk…

1,4-Dihydropyrazine-2,3-dione

Dodavatel: Apollo Scientific
Varování

OR21538-1G OR21538-250MG
APOSOR21538-1GEA 1820 CZK
APOSOR21538-1G APOSOR21538-250MG
1,4-Dihydropyrazine-2,3-dione
1,4-Dihydropyrazine-2,3-dione
Vzorec: C₄H₄N₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00275454
CAS číslo: 931-18-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR