2-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}acetonitrile
Dodavatel: Apollo Scientific
OR21495-10G
OR21495-1G
OR21495-5G
APOSOR21495-10GEA
9860
CZK
APOSOR21495-10G
APOSOR21495-1G
APOSOR21495-5G
2-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}acetonitrile
2-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}acetonitrile
Vzorec:
C₁₅H₁₁Cl₂NO MW: 292,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00067741 CAS číslo: 175135-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...