7-Hydroxynaphthalene-1-carbonitrile
Dodavatel: Apollo Scientific
OR2133-1G
OR2133-250MG
APOSOR2133-1GEA
7770
CZK
APOSOR2133-1G
APOSOR2133-250MG
7-Hydroxynaphthalene-1-carbonitrile
7-Hydroxynaphthalene-1-carbonitrile
Vzorec:
C₁₁H₇NO Storage Temperature: Ambient |
MDL Number:
MFCD03695467 CAS číslo: 19307-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...