3-Chloropyridazine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-Chloro-1,2-diazine hydrochloride
OR2124-1G
OR2124-5G
APOSOR2124-1GEA
2940
CZK
APOSOR2124-1G
APOSOR2124-5G
3-Chloropyridazine hydrochloride
3-Chloropyridazine hydrochloride
Vzorec:
C₄H₃ClN₂·ClH Storage Temperature: Ambient |
MDL Number:
MFCD12547058 CAS číslo: 856847-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...