Tisk…

1-[4-(Aminomethyl)benzyl]-1H-pyrazole hydrochloride, tech

Dodavatel: Apollo Scientific
Varování

OR2066-1G OR2066-250MG
APOSOR2066-1GEA 7440 CZK
APOSOR2066-1G APOSOR2066-250MG
1-[4-(Aminomethyl)benzyl]-1H-pyrazole hydrochloride, tech
1-[4-(Aminomethyl)benzyl]-1H-pyrazole hydrochloride
Vzorec: C₁₁H₁₃N₃·ClH
Storage Temperature: Ambient
MDL Number: MFCD07690519
CAS číslo: 904696-62-4
UN: 2811
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR