2-Bromo-6-(1-ethoxyethoxy)naphthalene 95+%
Dodavatel: Apollo Scientific
|
Varování
|
OR2046-1G
OR2046-5G
APOSOR2046-1GEA
4800
CZK
APOSOR2046-1G
APOSOR2046-5G
2-Bromo-6-(1-ethoxyethoxy)naphthalene 95+%
2-Bromo-6-(1-ethoxyethoxy)naphthalene
Vzorec:
C₁₄H₁₅BrO₂ MW: 295,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08741420 CAS číslo: 142524-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...