Tisk…

4-Propylphenylacetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-(4-Propylphenyl)acetic acid

OR2009-5G OR2009-25G OR2009-100G
APOSOR2009-5GEA 1330 CZK
APOSOR2009-5G APOSOR2009-25G APOSOR2009-100G
4-Propylphenylacetic acid
4-Propylphenylacetic acid
Vzorec: C₁₁H₁₄O₂
MW: 178.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00600398
CAS číslo: 26114-12-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR