Tisk…

4-Cyano-1H-indole-2-boronic acid, pinacol ester 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-4-carbonitrile

OR20047-1G OR20047-250MG
APOSOR20047-1GEA 8730 CZK
APOSOR20047-1G APOSOR20047-250MG
4-Cyano-1H-indole-2-boronic acid, pinacol ester 95%
4-Cyano-1H-indole-2-boronic acid, pinacol ester
Vzorec: C₁₅H₁₇BN₂O₂
MW: 268,119 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11858395
CAS číslo: 1256359-19-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR