Tisk…

2-Amino-3-nitrobenzamide

Dodavatel: Apollo Scientific

OR200158-1G OR200158-250MG OR200158-500MG OR200158-2.5G
APOSOR200158-1GEA 11540 CZK
APOSOR200158-1G APOSOR200158-250MG APOSOR200158-500MG APOSOR200158-2.5G
2-Amino-3-nitrobenzamide
2-Amino-3-nitrobenzamide
Vzorec: C₇H₇N₃O₃
MW: 181,149 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11215471
CAS číslo: 313279-12-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR