Tisk…

N-(Cyclopropylmethyl)-2-hydroxy-5-methylbenzamide

Dodavatel: Apollo Scientific

OR200155-1G OR200155-50MG OR200155-250MG OR200155-500MG
APOSOR200155-1GEA 20110 CZK
APOSOR200155-1G APOSOR200155-50MG APOSOR200155-250MG APOSOR200155-500MG
N-(Cyclopropylmethyl)-2-hydroxy-5-methylbenzamide
N-(Cyclopropylmethyl)-2-hydroxy-5-methylbenzamide
Vzorec: C₁₂H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD11130810
CAS číslo: 1019405-20-9

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR