5-Bromo-6-chloronicotinoyl chloride
Dodavatel: Apollo Scientific
OR200143-1G
OR200143-100MG
OR200143-250MG
OR200143-500MG
OR200143-2.5G
APOSOR200143-1GEA
16770
CZK
APOSOR200143-1G
APOSOR200143-100MG
APOSOR200143-250MG
APOSOR200143-500MG
APOSOR200143-2.5G
5-Bromo-6-chloronicotinoyl chloride
5-Bromo-6-chloronicotinoyl chloride
Vzorec:
C₆H₂BrCl₂NO Storage Temperature: Ambient |
MDL Number:
MFCD16037365 CAS číslo: 78686-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...