Tisk…

(2-Methoxyphenoxy)acetyl chloride

Dodavatel: Apollo Scientific

OR200141-1G OR200141-250MG OR200141-500MG
APOSOR200141-1GEA 14150 CZK
APOSOR200141-1G APOSOR200141-250MG APOSOR200141-500MG
(2-Methoxyphenoxy)acetyl chloride
(2-Methoxyphenoxy)acetyl chloride
Vzorec: C₉H₉ClO₃
MW: 200,619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02223230
CAS číslo: 40926-73-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR